Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrite Nitrogen Standard, 250 ppm N (821 ppm NO2-), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
16 oz.
Specifications
Specifications
Packaging | Natural Poly Bottle |
CAS Min % | 98.0, 0.13 |
CAS Max % | 100.0, 0.13 |
Boiling Point | 100°C |
Quantity | 16 oz. |
Chemical Name or Material | Nitrogen Standard, 1 mL = 0.25 mg N as NO2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction