Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonia Nitrogen Standard, 1000 ppm N (1216 ppm NH3), Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
CAS Min % | 98.0, 0.37 |
CAS Max % | 100.0, 0.39 |
Methods | APHA/ASTM/EPA for Ammonia Nitrogen |
Equivalence | 1mL = 1mg N |
Boiling Point | 100°C |
Quantity | 500 mL |
Concentration or Composition (by Analyte or Components) | 1000ppm N, 1216ppm NH3 |
Chemical Name or Material | Nitrogen Standard (as Ammonia) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction