Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrogen Standard, as Ammonia, 1mL = 1mg N, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 54554

Packaging Poly Bottle
Quantity 120mL

 View more versions of this product

Catalog No. 5455-4

  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7732-18-5,12125-02-9
98.0,0.37
100.0,0.39
Nitrogen Standard (as Ammonia)

Specifications

7732-18-5,12125-02-9
100.0,0.39
Poly Bottle
1mL = 1mg N
Nitrogen Standard (as Ammonia)
98.0,0.37
1000ppm N, 1216ppm NH3
APHA/ASTM/EPA for Ammonia Nitrogen
120mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit