Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrite Nitrogen Standard, 1000 ppm N (3285 ppm NO2-), Ricca Chemical
SDP

Catalog No. 546116
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7758-09-0, 7732-18-5
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 0.62
CAS Max % 100.0, 0.65
Analyte or Component Names KNO2- in water
Equivalence 1 mL = 1 mg N as NO2
pH Approximately 7
Boiling Point 100°C
Element Multi-element
Solvent or Matrix Water
Quantity 500 mL
Grade ASTM
Concentration or Composition (by Analyte or Components) 1000ppm N, 3285ppm NO2-
Chemical Name or Material Nitrogen Standard (as Nitrite)
Show More Show Less
ShelfLife 6 month shelf life refrigerated.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.