Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane 98.0+%, TCI America™
SDP

Catalog No. N0209100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0209100G 100 g
N020925G 25 g
N0209500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0209100G Supplier TCI America Supplier No. N0209100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 101°C
Quantity 100 g
UN Number 1261
Formula Weight 61.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nitromethane
EINECSNumber (2)-0191
RTECSNumber PA9800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.