Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitronium tetrafluoroborate, 0.5M solution in sulfolane

Catalog No. AC175132000
Click to view available options
Quantity:
25 mL
200 mL
Packaging:
Glass bottle

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitronium tetrafluoroborate
Color Colorless through Yellow
CAS 13826-86-3, 126-33-0
CAS Min % 5.03, 94.24
CAS Max % 5.76, 94.97
Physical Form Solution
Quantity 200 mL
Molecular Formula BF4NO2
Linear Formula NO2BF4
MDL Number MFCD00011432
Fieser 01,742; 04,358; 05,476; 08,361; 09,324; 06,520; 07,324; 09,414; 12,435; 13,274; 14,282; 17,314
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
Solubility Information Solubility in water: reacts
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES [O-][NH+]=O.F[B-](F)(F)F
IUPAC Name nitronium;tetrafluoroborate
Molecular Weight (g/mol) 132.81
PubChem CID 11073463
Formula Weight 132.81
Packaging Glass bottle
Specific Gravity 1.268
Density 1.2680g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.