Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitronium tetrafluoroborate, 97%

Catalog No. AC174380250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
PubChem CID 11073463
IUPAC Name nitronium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point 200.0°C
Color Colorless through White
Physical Form Adhering Crystals
Quantity 25 g
Assay Percent Range 96% min
Linear Formula NO2BF4
Fieser 01,742; 04,358; 05,476; 08,361; 09,324; 01,747; 04,360; 07,253; 08,364; 14,215
Solubility Information Solubility in water: may decompose
Formula Weight 132.81
Percent Purity 97%
Packaging Plastic bottle
Chemical Name or Material Nitronium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 237-533-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.