Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nitrosobenzene

Catalog No. N2460910G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N2460910G 10 g
N2460925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N2460910G Supplier Sigma Aldrich Supplier No. N2460910G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nitrosobenzene

Specifications

CAS 586-96-9
Melting Point 65°C to 69°C (lit.)
Boiling Point 59°C (18 mmHg)
Molecular Formula C6H5NO
Linear Formula C6H5NO
MDL Number MFCD00002059
Quantity 10 g
Synonym NOB
Molecular Weight (g/mol) 107.11
Percent Purity ≥97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.