Learn More
Nitrotryptophan Antibody (4F8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259355AF647
Description
Nitrotryptophan Monoclonal specifically detects Nitrotryptophan in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Nitrotryptophan | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for 6-Nitrotyrptophan modified proteins. Doss not cross-react with 7-Ketocholoesterol modified proteins. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
4F8 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |