Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Antibody (39B6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11096877H
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Rat, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Nitrotyrosine | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
Primary | |
Rat, Non-species specific | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
39B6 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Nitrotyrosine | |
Hybridoma line 39B6 | |
0.1 mL | |
Recognizes 3-nitrotyrosine moieties. No detectable cross-reactivity with non-nitrated tyrosine. Not species specific. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction