Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Antibody (EM-30), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196130C
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Human, Mouse, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Nitrotyrosine | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
EM-30 | |
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Nitrotyrosine | |
NO2-Tyr-CH2-Thyroglobulin | |
0.1 mL | |
Primary | |
Human, Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction