Learn More
Nitrotyrosine Antibody (EM-30), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP196130IR
Catalog No. NBP196130IR
Only null left
Add to Cart
Description
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Human, Mouse, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Nitrotyrosine
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein A or G purified
RUO
The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
EM-30
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Nitrotyrosine
NO2-Tyr-CH2-Thyroglobulin
0.1 mL
Primary
Human, Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.