Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotyrosine Mouse anti-Human, Mouse, Rat, APC, Clone:39B6, Abnova™

Catalog No. 89333692 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-692 Supplier Abnova Corporation Supplier No. MAB16962
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Nitrotyrosine.

Specifications

Antigen Nitrotyrosine
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 39B6
Conjugate APC
Dilution Antibody Microarray
ELISA
Flow Cytometry
Immunocytochemistry
Immunofluorescence
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100)
Immunoprecipitation
Western Blot (1:1400)
The optimal working dilution sho
Formulation In PBS (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen Hybridoma line 39B6.
Quantity 100 μg
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.