Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$205.50 - $498.16
Specifications
| Antigen | Nitrotyrosine |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
Nitrotyrosine Polyclonal antibody specifically detects Nitrotyrosine in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Nitrotyrosine | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Nitrotyrosine | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Lipid and Metabolism, Neuroscience, Neurotransmission, Tyrosine Kinases | |
| PBS, 50% glycerol, pH7.3 | |
| A synthetic peptide corresponding to a sequence containing nitrated Y | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title