Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nizatidine, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 g
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 76963-41-2 |
| Physical Form | Neat |
| Linear Formula | C12H21N5O2S2 |
| MDL Number | MFCD00865660 |
| Synonym | N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N′-methyl-2-nitro-1,1-ethenediamine |
| Type | Analytical Standard |
| Quantity | 5 g |
| Formula Weight | 331.46 |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction