Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Njmu-R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544812 Supplier Thermo Scientific Supplier No. PA544812
Add to Cart
Add to Cart

Njmu-R1 Polyclonal Antibody

Njmu-R1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Njmu-R1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C17orf75
Gene Accession No. Q9HAS0
Gene Alias 5730455P16Rik; C17orf75; chromosome 17 open reading frame 75; NJMU-R1; Protein Njmu-R1; RGD1310429; RIKEN cDNA 5730455P16 gene; sensitization to ricin complex subunit 2; similar to Protein Njmu-R1; spermatogenesis-related protein; SRI2
Gene Symbols C17orf75
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C17orf75 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64149
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.