Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Njmu-R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552046 Supplier Invitrogen™ Supplier No. PA552046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSSHYCLYS YRGSRLAQQR GDSEDGSPSG TNAETPSGDD FSLSLADTNL PSEVEPELRS FIAKRLSRGA VFEGLGNVAS VELKIPGYRV GCYYCLFQNE KLLPETVTI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

As component of the WDR11 complex acts together with TBC1D23 to facilitate the golgin-mediated capture of vesicles generated using AP-1 (PubMed:29426865). May have a role in spermatogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Njmu-R1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C17orf75
Gene Accession No. Q9CYI0, Q9HAS0
Gene Alias 5730455P16Rik; C17orf75; chromosome 17 open reading frame 75; NJMU-R1; Protein Njmu-R1; RGD1310429; RIKEN cDNA 5730455P16 gene; sensitization to ricin complex subunit 2; similar to Protein Njmu-R1; spermatogenesis-related protein; SRI2
Gene Symbols 5730455P16Rik, C17orf75
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Njmu-R1. Recombinant protein control fragment (Product #RP-102398).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64149, 70591
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.