Learn More
Invitrogen™ Njmu-R1 Polyclonal Antibody

Description
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Njmu-R1 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | C17orf75 |
| Gene Accession No. | Q9CYI0, Q9HAS0 |
| Gene Alias | 5730455P16Rik; C17orf75; chromosome 17 open reading frame 75; NJMU-R1; Protein Njmu-R1; RGD1310429; RIKEN cDNA 5730455P16 gene; sensitization to ricin complex subunit 2; similar to Protein Njmu-R1; spermatogenesis-related protein; SRI2 |
| Gene Symbols | 5730455P16Rik, C17orf75 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.