Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK-1.1 Mouse anti-Mouse, APC, Clone: PK136, BD

Catalog No. BDB550627 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550627 0.1 mg
BDB561117 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB550627 Supplier BD Biosciences Supplier No. 550627
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In the mouse, at least three members of the Klrb (Killer cell lectin-like receptor, subfamily b; formerly NKR-P1) gene family have been identified (Klrb1a/NKR-P1A, Klrb1b/NKR-P1B, and Klrb1c/NKR-P1C); but in the human gene family, a single homologue has b

Host Species: Mouse
Clone: PK136
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse NK-1+ Spleen and Bone Marrow

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen NK-1.1
Applications Flow Cytometry
Classification Monoclonal
Clone PK136
Concentration 0.2mg/mL
Conjugate APC
Description Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse NK-1+ Spleen and Bone Marrow
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.