Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK-1.1 Mouse anti-Mouse, BV711, Clone: PK136, BD Horizon™

Catalog No. BDB569723 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569723 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569723 Supplier BD Biosciences Supplier No. 569723
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NK-1.1 Monoclonal antibody specifically detects NK-1.1 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NK-1.1
Applications Flow Cytometry
Classification Monoclonal
Clone PK136
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c
Host Species Mouse
Immunogen Mouse NK-1+ Spleen and Bone Marrow Cells
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17059,80782
Target Species Mouse
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.