Learn More
NK-1.1 Mouse anti-Mouse, FITC, Clone: PK136, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 553164
Description
In the mouse, at least three members of the Klrb (Killer cell lectin-like receptor, subfamily b; formerly NKR-P1) gene family have been identified (Klrb1a/NKR-P1A, Klrb1b/NKR-P1B, and Klrb1c/NKR-P1C); but in the human gene family, a single homologue has b
Host Species: Mouse
Clone: PK136
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse NK-1+ Spleen and Bone Marrow
Flow Cytometry

Specifications
NK-1.1 | |
Monoclonal | |
0.5mg/mL | |
Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
PK136 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse NK-1+ Spleen and Bone Marrow | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.