Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NK1.1, Azide Free, Mouse, Unlabeled, Clone: PK136,

Catalog No. MABF1495ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1495ZMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1495ZMI Supplier MilliporeSigma™ Supplier No. MABF1495Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The NK cell receptor protein 1 (NKR-P1), a.k.a. CD161, molecules represent a family of type II transmembrane C-type lectin-like receptors expressed predominantly by NK cells. A total of five murine NKR-P1-coding genes (Klrb1a/b/c/d/f) have been identified, with NKR-P1A/C/F (CD161a/c/f) functioning as stimulatory receptors that possess a charged transmembrane arginine esidue thought to be important for association with the FcRγ adaptor protein, and NKR-P1B/D as two inhibitory receptors that possess a consensus cytoplasmic ITIM (L/VxYxxL/I/V) motif. The ITIM motif is shown to mediate NKR-P1B recruitment of Src homology 2 (SH2)-containing protein tyrosine phosphatase-1 (SHP-1) in a phosphorylation-dependent manner. All murine NKR-P1 proteins possess a Cys-X-Cys-Pro (CxCP) motif that mediates association with the Src-related nonreceptor protein tyrosine kinase p56lck. Both NKR-P1B and NKR-P1C functionally associate with p56lck, while mutation of the CxCP motif abolishes NKR-P1B/C-mediated signal transduction. Likewise, mutation of ITIM motif abolishes NKR-P1B SHP-1 association as well as NKR-P1B-dependent inhibitory signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NK1.1, Azide Free
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone PK136
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in PBS without preservatives.
Gene Klrb1b, Ly55b, Nkrp1b, Klrb1c, Ly55c, Nkrp1c
Gene Accession No. P27812;P27814
Host Species Mouse
Immunogen Mouse spleen and bone marrow cells (Koo, G.C., and Peppard, J.R. (1984). Hybridoma. 3(3):301-303).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) ABB72026;NP_001153376
Test Specificity Clone PK136 recognizes murine NKR-P1B/CD161b (UniProt P27812) and NK1.1/NKR-P1C/CD161c (UniProt P27814), but not CD161a/NKR-P1A (P27811). Upon activation by PK136 binding, NKR-P1C/NK1.1 signals to activate, while NKR-P1B signals to suppress, NK cells for cytotoxic killing of target cells (Carlyle, J.R., et al. (1999). J. Immunol. 162(10):5917-5923). Epitope is present in both spliced isoforms of murine NK1.1/CD161c reported by UniProt (P27814). Clone PK136 does not react with CD161a (by Klrb1a gene) or C57BL/6 mouse strain-derived CDC161b variant (by Klrb1b gene, also known as Klrb1d). There exists a fifth murine CD161 gene Klrb1f, reactivity of this mAb toward this gene product has not been established.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.