Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NK1R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NK1R |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NK1R Polyclonal specifically detects NK1R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NK1R | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
6869 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FFLLPYINPDLYLKKFIQQVY | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
TACR1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title