Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NK1R Antibody, PE/Cy5.5, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300119PECY55
Description
NK1R Polyclonal specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
NK1R | |
Polyclonal | |
Flow Cytometry | |
NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
Rabbit | |
Affinity Purified | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
6869.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
TACR1 | |
A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor. [UniProt No. P25103] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction