Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKAIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552355 Supplier Invitrogen™ Supplier No. PA552355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRDFIMTFNT SLHRSWWMEN GPGCLVTPVL NSRLALEDHH VISVTGCLLD YPY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

NKAIN1 is a member of a family of mammalian proteins with similarity to Drosophila Nkain and interacts with the beta subunit of Na,K-ATPase (ATP1B1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKAIN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NKAIN1
Gene Accession No. Q4KMZ8
Gene Alias FAM77C; family with sequence similarity 77, member C; Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 1; Na+/K+ transporting ATPase interacting 1; NKAIN1; Protein FAM77C; Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 1
Gene Symbols NKAIN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NKAIN1. Recombinant protein control fragment (Product #RP-89706).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.