Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578578 Supplier Invitrogen™ Supplier No. PA578578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Mouse (98%), Rat (97%), Zebrafish (82%), Xenopus laevis (85%), Pig (97%), Chicken (91%), Rhesus Monkey (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NKAP is a transcriptional repressor that associates with the NOTCH (see MIM 190198) corepressor complex and is required for T-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKAP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene NKAP
Gene Accession No. Q8N5F7
Gene Alias 2610020O08Rik; AA987160; AI849147; AL024236; c6orf194l; FLJ22626; FLJ22626-like; NF-kappaB activating protein; NF-kappa-B-activating protein; NF-kappa-B-activating protein; LOW QUALITY PROTEIN: NF-kappa-B-activating protein; NFKB activating protein; NKAP; nkapl; nuclear NF-kappaB activating protein; wu:fj36d09; wu:fj49c09; zgc:136458
Gene Symbols NKAP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human NKAP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79576
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.