Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115654 Supplier Invitrogen™ Supplier No. PA5115654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NKAP.

NKAP is a transcriptional repressor that associates with the NOTCH (see MIM 190198) corepressor complex and is required for T-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKAP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NKAP
Gene Accession No. Q4V7C9, Q8N5F7, Q9D0F4
Gene Alias 2610020O08Rik; AA987160; AI849147; AL024236; c6orf194l; FLJ22626; FLJ22626-like; NF-kappaB activating protein; NF-kappa-B-activating protein; NF-kappa-B-activating protein; LOW QUALITY PROTEIN: NF-kappa-B-activating protein; NFKB activating protein; NKAP; nkapl; nuclear NF-kappaB activating protein; wu:fj36d09; wu:fj49c09; zgc:136458
Gene Symbols NKAP
Host Species Rabbit
Immunogen A synthesized peptide derived from human NKAP(Accession Q8N5F7), corresponding to amino acid residues G6-L56.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298342, 67050, 79576
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.