Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKAPD1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299180PE
Description
NKAPD1 Polyclonal antibody specifically detects NKAPD1 in Human samples. It is validated for Western BlotSpecifications
NKAPD1 | |
Polyclonal | |
Western Blot | |
C11orf57, chromosome 11 open reading frame 57, FLJ10726, hypothetical protein LOC55216 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human NKAPD1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55216 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction