Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKAPL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NKAPL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NKAPL Polyclonal specifically detects NKAPL in Human samples. It is validated for Western Blot.Specifications
NKAPL | |
Polyclonal | |
Rabbit | |
MGC126734, NFKB activating protein-like | |
NKAPL | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
222698 | |
Synthetic peptides corresponding to NKAPL (NFKB activating protein-like) The peptide sequence was selected from the middle region of NKAPL)(50ug). Peptide sequence KTSRSRNKKKRKNKSSKRKHRKYSDSDSNSESDTNSDSDDDKKRVKAKKK. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title