Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NKAPL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598478
Description
NKAPL Polyclonal Antibody for Western Blot, ELISA
The function of NKAPL remains unknown.
Specifications
NKAPL | |
Polyclonal | |
Unconjugated | |
NKAPL | |
bA424I5.1; C6orf194; NFKB activating protein like; NFKB activating protein-like; NKAPL; NKAP-like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
222698 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q5M9Q1 | |
NKAPL | |
Peptide sequence from Human NKAP-like protein (114-123AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction