Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561125 Supplier Invitrogen™ Supplier No. PA561125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAVCKEPQEE VVPGGGRSKR DPDLYQLLQR LFKSHSSLEG LLKALSQAST DPKESTSPEK RDMHDFFVGL M Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 59%.

proNeurokinin B (proNKB) is a neuropeptide belonging to the Tachykinin family, widely expressed in the Central Nervous System (CNS). It plays a neuromodulatory role in various brain functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAC3
Gene Accession No. P55099, Q9UHF0
Gene Alias gamma tachykinin 3; HH10; neurokinin 2; neurokinin A; neurokinin alpha; neurokinin b; neurokinin B-like protein Zneurok1; Neurokinin-B; neuromedin K; neuromedin L; Neuromedin-K; neuropeptide K; NKB; NKNB; PPT-B; preprotachykinin B; preprotachykinin-B; PRO1155; substance K; Tac2; Tac3; tachykinin 2; tachykinin 3; tachykinin-3; UNQ585/PRO1155; ZNEUROK1
Gene Symbols Tac2, TAC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NKB. Recombinant protein control fragment (Product #RP-97814).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21334, 6866
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.