Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA132353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132353 Supplier Invitrogen™ Supplier No. PA132353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

proNeurokinin B (proNKB) is a neuropeptide belonging to the Tachykinin family, widely expressed in the Central Nervous System (CNS). It plays a neuromodulatory role in various brain functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKB
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TAC3
Gene Accession No. P08435, P55099, Q9UHF0
Gene Alias gamma tachykinin 3; HH10; neurokinin 2; neurokinin A; neurokinin alpha; neurokinin b; neurokinin B-like protein Zneurok1; Neurokinin-B; neuromedin K; neuromedin L; Neuromedin-K; neuropeptide K; NKB; NKNB; PPT-B; preprotachykinin B; preprotachykinin-B; PRO1155; substance K; Tac2; Tac3; tachykinin 2; tachykinin 3; tachykinin-3; UNQ585/PRO1155; ZNEUROK1
Gene Symbols Tac2, TAC3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of mouse proNeurokinin B (~93% homology to the rat sequence).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21334, 29191, 6866
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.