Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980038
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0038 Supplier Bioss Supplier No. BS0070R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

proNeurokinin B (proNKB) is a neuropeptide belonging to the Tachykinin family, widely expressed in the Central Nervous System (CNS). It plays a neuromodulatory role in various brain functions.

Specifications

Antigen NKB
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TAC3
Gene Accession No. Q9UHF0
Gene Alias gamma tachykinin 3; HH10; neurokinin 2; neurokinin A; neurokinin alpha; neurokinin b; Neurokinin B-like protein Zneurok1; Neurokinin-B; neuromedin K; neuromedin L; Neuromedin-K; neuropeptide K; NKB; NKNB; PPT-B; preprotachykinin B; Preprotachykinin-B; PRO1155; substance K; Tac2; TAC3; tachykinin 2; tachykinin 3; Tachykinin-3; UNQ585/PRO1155; ZNEUROK1
Gene Symbols TAC3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NKB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6866
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.