Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NKCC1/2, Mouse, Unlabeled, Clone: T4,

Catalog No. MABS1237MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1237MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1237MI Supplier MilliporeSigma™ Supplier No. MABS1237
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Solute carrier family 12 member 1 (UniProt Q13621; a.k.a. Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2) and solute carrier family 12 member 2 (UniProt P55011; a.k.a. Basolateral Na-K-Cl symporter, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1) are encoded by the SLC12A1 (a.k.a. NKCC2; Gene ID 6557) and SLC12A2 (a.k.a. NKCC1; Gene ID 6558) gene, respectively, in human. NKCC1 (SLC12A2) and NKCC2 (SLC12A1) belong to the subfamily of cation-chloride cotransporters (CCCs) that mediate electroneutral transport of sodium, potassium and chloride across the plasma membrane and are inhibited by bumetanide and furosemide-type of compounds. NKCCs play an important role in cell volume regulation, where NKCC2 is involved in reabsorption of salt and osmotically-obliged water in renal epithelial cells, while NKCC1 has a major impact on intracellular chloride ion concentration ([Cl-]i) in vascular smooth muscle cells (VSMCs) and neuronal cells. In primary hypertension, NKCC1 activity is increased in VSMCs and paraventricular nucleus (PVN) neurons, leading to elevated peripheral resistance in systemic circulation and activation of sympathetic nerve system (SNS), respectively. Human NKCC1 and NKCC2 are 12-transmembrane proteins having their N- and C-terminal ends exposed at the cytoplasmic side (a.a. 1-285 and 921-1212 of NKCC1; a.a. 1-177 and 814-1099 of NKCC2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKCC1/2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone T4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene SLC12A1, SLC12A2, NKCC2, NKCC1
Gene Accession No. P55011
Host Species Mouse
Immunogen Recombinant human NKCC1 C-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001037.1
Test Specificity Clone T4 recognizes both NKCC1 and NKCC2 by targeting a C-terminal region epitope exposed only under denaturing condition, and unaccessible for antibody binding when NKCC assumes its native conformation (Lytle, C., et al. (1995). Am. J. Physiol. 269(6 Pt 1):C1469-C1505).
Target Species Avian
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.