Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NKCC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569256
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
NKCC1 mediates sodium and chloride transport and reabsorption. NKCC1 is a membrane protein and is important in maintaining proper ionic balance and cell volume. NKCC1 is an electrically silent transporter system that mediates sodium and chloride reabsorption. Further, NKCC1 plays a vital role in the regulation of ionic balance and cell volume and is phosphorylated in response to DNA damage. Three transcript variants encoding two different isoforms have been found for NKCC1. Diseases associated with SLC12A2 include Basosquamous Carcinoma and Metatypical Basal Cell Carcinoma.
Specifications
NKCC1 | |
Polyclonal | |
Unconjugated | |
SLC12A2 | |
9330166H04Rik; basolateral NaK(2Cl) cotransporter; basolateral Na-K-Cl symporter; BSC; BSC2; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; little ears; lte; mBSC2; Na-K-2Cl- cotransporter; Na-K-Cl cotransporter; NKCC1; PPP1R141; protein phosphatase 1, regulatory subunit 141; Slc12a2; slc12a4; sodium/potassium/chloride cotransporters; sodium/potassium/chloride transporters; solute carrier family 12 (potassium/chloride transporters), member 2; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 (sodium/potassium/chloride transporter), member 2; solute carrier family 12 (sodium/potassium/chloride transporters), member 2; solute carrier family 12 member 2; solute carrier family 12, member 2; sy-ns; zfnkcc1; zgc:85961; zmp:0000000537 | |
Rabbit | |
Protein A | |
RUO | |
6558 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P55011 | |
SLC12A2 | |
synthetic peptide directed towards the C terminal region of human NKCC1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction