Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKCC1/SLC12A2 Antibody (915911), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8528H
Description
NKCC1/SLC12A2 Monoclonal antibody specifically detects NKCC1/SLC12A2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| NKCC1/SLC12A2 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6558 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 915911 | |
| Flow Cytometry, CyTOF-ready | |
| Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2 | |
| NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2, Met1-Ser1212, Accession # P55011 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction