Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKCC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541995 Supplier Invitrogen™ Supplier No. PA541995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NEKKSRGFFN YQASIFAENF GPRFTKGEGF FSVFAIFFPA ATGILAGANI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKCC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC12A1
Gene Accession No. Q13621
Gene Alias AI788571; apical Na(2Cl)K cotransporter; BSC1; bumetanide-sensitive cotransporter type 1; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform A; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform B; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform F; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; D630042G03Rik; Kidney-specific Na-K-Cl symporter; mBSC1; MGC48843; Na-K-2Cl cotransporter; NKCC2; NKCC2A variant A; SLC12A1; solute carrier family 12 (sodium/potassium/chloride transporter), member 1; solute carrier family 12 (sodium/potassium/chloride transporters), member 1; solute carrier family 12 member 1; Solute carrier family 12 member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); solute carrier family 12, member 1; Solute carrier family 12, member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); urehr3
Gene Symbols SLC12A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC12A1 (aa 351-400).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6557
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.