Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKCC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553263 Supplier Invitrogen™ Supplier No. PA553263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLNNSSNVF LDSVPSNTNR FQVSVINENH ESSAAADDNT DPPHYEETSF GDEAQKRLRI SFRPGNQECY DNFLQSGETA KTDASFHAYD SHTNTYYLQT FGHNTMDAVP KIEYYRNTGS I Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKCC2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC12A1
Gene Accession No. Q13621
Gene Alias AI788571; apical Na(2Cl)K cotransporter; BSC1; bumetanide-sensitive cotransporter type 1; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform A; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform B; bumetanide-sensitive Na-K-Cl cotransport protein splice isoform F; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; D630042G03Rik; Kidney-specific Na-K-Cl symporter; mBSC1; MGC48843; Na-K-2Cl cotransporter; NKCC2; NKCC2A variant A; SLC12A1; solute carrier family 12 (sodium/potassium/chloride transporter), member 1; solute carrier family 12 (sodium/potassium/chloride transporters), member 1; solute carrier family 12 member 1; Solute carrier family 12 member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); solute carrier family 12, member 1; Solute carrier family 12, member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); urehr3
Gene Symbols SLC12A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC12A1. Recombinant protein control fragment (Product #RP-91245).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6557
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.