Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKCC2/SLC12A1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298480MFV500
Description
NKCC2/SLC12A1 Polyclonal antibody specifically detects NKCC2/SLC12A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
NKCC2/SLC12A1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
BSC1, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2, Kidney-specific Na-K-Cl symporter, MGC48843, NKCC2A variant A, NKCC2Na-K-2Cl cotransporter, solute carrier family 12 (sodium/potassium/chloride transporters), member 1, solute carrier family 12 member 1 | |
Produced in rabbits immunized with E. coli-derived Human NKCC2/SLC12A1 fragment. | |
0.1 mL | |
ABC Transporters, Cancer, Neuroscience | |
6557 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction