Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2A/C/E Rat anti-Mouse, APC, Clone: 20d5, BD Pharmingen™

Catalog No. BDB564383 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564383 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564383 Supplier BD Biosciences Supplier No. 564383

Rat Monoclonal Antibody

NKG2A/C/E Monoclonal antibody specifically detects NKG2A/C/E in Mouse samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKG2A/C/E
Applications Flow Cytometry
Classification Monoclonal
Clone 20d5
Concentration 0.2mg/mL
Conjugate APC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias NKG2A/NKG2C/NKG2E; Klrc1/Klrc2/Klrc3; CD159a/CD159c/CD159e; CD159
Host Species Rat
Immunogen Mouse CD94 and NKG2A Transfected Cell Line
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions and unconjugated antibody and free APC were removed.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.