Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKG2A/CD159a/KLRC1 Antibody (131411R) [DyLight 550], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1059RL
Description
NKG2A/CD159a/KLRC1 Monoclonal antibody specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
NKG2A/CD159a/KLRC1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
131411R | |
Flow Cytometry, CyTOF-ready | |
CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
BaF3 mouse pro-B cell line transfected with human NKG2A and CD94 | |
0.1 mL | |
Adaptive Immunity, Immunology, Signal Transduction | |
3821 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction