Learn More
NKG2A (CD159a) Mouse anti-Human, BUV737, Clone: 131411, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | NKG2A (CD159a) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 131411 |
| Conjugate | Brilliant Ultraviolet 737 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | KLRC1; CD159A; NKG2A; NK cell receptor A; natural killer cell lectin; natural killer group protein 2; C-lectin type II protein |
| Host Species | Mouse |
| Immunogen | Human NKG2A and CD94 Transfected Cell Line |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.