Learn More
Invitrogen™ NKG2A/NKG2C/NKG2E Monoclonal Antibody (20d5), FITC

Description
NKG2A/NKG2C/NKG2E Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | NKG2A/NKG2C/NKG2E, NKG2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 20d5 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | Klrc1 |
| Gene Accession No. | 0 |
| Gene Alias | CD159a; cd159c; cd159e; Cluster of Differentiation 159a; killer cell lectin-like receptor subfamily C, member 1; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; Klrc1; Klrc2; Klrc3; natural killer cell group 2E cell receptor; NKG2; NKG2A; NKG2-A/NKG2-B type II integral membrane protein; NKG2B; NKG2C; Nkg2e |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.