Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NKG2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521949
Description
Recommended positive controls: A431, HepG2. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
Specifications
NKG2A | |
Polyclonal | |
Unconjugated | |
Klrc1 | |
CD159 antigen-like family member A; CD159A; C-lectin type II protein; Cluster of Differentiation 159a; killer cell lectin like receptor C1; killer cell lectin-like receptor subfamily C, member 1; Klrc1; natural killer cell lectin; natural killer group protein 2; NK cell receptor A; NKG2; NKG2-1/B activating NK receptor; NKG2A; NKG2-A/B type II integral membrane protein; NKG2-A/B-activating NK receptor; NKG2-A/NKG2-B type II integral membrane protein; NKG2B; rNKG2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3821 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P26715 | |
Klrc1 | |
Synthetic peptide corresponding to a region within amino acids 1 and 45 of KLRC1 (Uniprot ID#P26715). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction