Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NKG2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588485
Description
Immunogen sequence: RHCGHCPEEW ITYSNSCYYI GKERRTWEES LLACTSKNSS LLSIDNEEEM KFLSIISPSS WIGVFRNSSH HPWVTMNGLA FKHEIKDSDN AELNCAVLQV NRLKSAQCGS SIIYHCKHKL; Positive Samples: Mouse testis, Mouse liver; Cellular Location: Membrane, Single-pass type II membrane protein.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
Specifications
NKG2A | |
Polyclonal | |
Unconjugated | |
Klrc1 | |
Klrc1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 96-215 of human KLRC1 (NP_0155671). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD159 antigen-like family member A; CD159A; C-lectin type II protein; Cluster of Differentiation 159a; killer cell lectin like receptor C1; killer cell lectin-like receptor subfamily C, member 1; Klrc1; natural killer cell lectin; natural killer group protein 2; NK cell receptor A; NKG2; NKG2-1/B activating NK receptor; NKG2A; NKG2-A/B type II integral membrane protein; NKG2-A/B-activating NK receptor; NKG2-A/NKG2-B type II integral membrane protein; NKG2B; rNKG2A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16641 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction