Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528836 Supplier Thermo Scientific Supplier No. PA528836

Rabbit Polyclonal Antibody

Recommended positive controls: K562. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKG2A
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Klrc1
Gene Accession No. P26715
Gene Alias CD159 antigen-like family member A; CD159A; C-lectin type II protein; Cluster of Differentiation 159a; killer cell lectin like receptor C1; killer cell lectin-like receptor subfamily C, member 1; Klrc1; natural killer cell lectin; natural killer group protein 2; NK cell receptor A; NKG2; NKG2-1/B activating NK receptor; NKG2A; NKG2-A/B type II integral membrane protein; NKG2-A/B-activating NK receptor; NKG2-A/NKG2-B type II integral membrane protein; NKG2B; rNKG2A
Gene Symbols Klrc1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 171 and 233 of Human KLRC1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3821
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.