Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKG2C/CD159c/KLRC2 Antibody (134591), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB138H
Description
NKG2C/CD159c/KLRC2 Monoclonal antibody specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry, CyTOF-reportedSpecifications
| NKG2C/CD159c/KLRC2 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 3822 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 134591 | |
| Flow Cytometry, CyTOF-reported | |
| CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
| BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction