Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NKG2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566127
Description
Immunogen sequence: CAVLQVNRLKS AQCGSSMIYH CKH Highest antigen sequence identity to the following orthologs - mouse 38%, rat 38%.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
Specifications
NKG2C | |
Polyclonal | |
Unconjugated | |
KLRC2 | |
CD159 antigen-like family member C; CD159A; CD159c; cd159e; killer cell lectin like receptor C2; killer cell lectin-like receptor subfamily C, member 2; KLRC1; KLRC2; NK cell receptor C; NKG2; NKG2A; NKG2C; NKG2-C; NKG2-C type II integral membrane protein; NKG2-C-activating NK receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P26717 | |
KLRC2 | |
Recombinant Human NKG2C. Recombinant protein control fragment (Product #RP-105632). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction