Learn More
NKG2D/CD314 Antibody (149810), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB139MFV610
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, Costimulation of T Cells, CyTOFSpecifications
| NKG2D/CD314 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Neuroscience, Signal Transduction, Stem Cells | |
| 22914 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Neutralization, T-Cell Activation, CyTOF | |
| 149810 | |
| Western Blot, Flow Cytometry, Neutralization, Costimulation of T Cells, CyTOF-ready | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| BaF3 mouse pro-B cell line transfected with human NKG2D/CD314 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |