Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKG2D/CD314 Antibody (1D11), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065956UV
Description
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
NKG2D/CD314 | |
Monoclonal | |
DyLight 350 | |
P26718 | |
KLRK1 | |
NKL cells. | |
0.25 mL | |
Primary | |
NB100-65956 is specific for CD314, also known as natural killer receptor G2 (NKG2D) and as killer cell lectin-like receptor subfamily K, member 1 (KLRK1). CD314 is a C-type lectin-like activating receptor which is expressed on most natural killer (NK) cells, CD8 T cells and gamma delta T cells. CD314 forms homodimers that signal through an associated DAP10 adaptor protein. Ligands of CD314 include MICA, MICB and UL16 binding protein (ULBP), which are inducibly expressed. Ligand binding to CD314 results in NK cell activation and potent costimulation of effector T cells. Clone 1D11 is reported to inhibit T cell recognition of MICA (1,6). | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1D11 | |
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
Mouse | |
Protein G purified | |
RUO | |
22914 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction